missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Monoclonal Antibody (OX-34), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517489
Additional Details : Weight : 0.01000kg
Description
CD2 Monoclonal antibody specifically detects CD2 in Rat samples. It is validated for Flow Cytometry.Specifications
CD2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P08921 | |
CD2 | |
T blasts prepared in mixed lymphocyte reactions with purified rat T helper cells against irradiated spleen. | |
200 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry | |
OX-34 | |
PE | |
CD2 | |
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
Mouse | |
Protein G | |
RUO | |
497761 | |
4° C, do not freeze | |
Liquid |