missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD200 Monoclonal Antibody (325516), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524048
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant mouse CD200 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow Cytometry.Specifications
CD200 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P41217 | |
Cd200 | |
Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
325516 | |
Unconjugated | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Mouse | |
Protein A/G | |
RUO | |
4345 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |