missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Monoclonal Antibody (LB21), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516766
2683.46 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD21 Monoclonal antibody specifically detects CD21 in Bovine, Feline, Goat, Human, Mink, Ovine samples. It is validated for Flow Cytometry.Specifications
CD21 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P20023 | |
CR2 | |
Human IM9 cell line. | |
100 μg | |
Primary | |
Bovine, Feline, Goat, Human, Mink, Ovine | |
Antibody | |
IgG1 |
Flow Cytometry | |
LB21 | |
FITC | |
CR2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
101094393, 102186615, 1380, 407126, 443017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |