missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD21 Monoclonal Antibody (21A/5), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517707
Additional Details : Weight : 0.01000kg
Description
MA5-17707 targets CD21/CR2 in FACS applications and shows reactivity with Human samples. The MA5-17707 immunogen is mouse monoclonal antibodies raised against human CD21.
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Flow Cytometry.Specifications
CD21 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P20023 | |
CR2 | |
Mouse monoclonal antibodies raised against human CD21. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
21A/5 | |
APC | |
CR2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
Mouse | |
Protein G | |
RUO | |
1380 | |
4° C, do not freeze | |
Liquid |