Learn More
CD22 Monoclonal Antibody (308501), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524023
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, -6, -7, -9, -10, -11, or recombinant mouse Siglec-F is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CD22 Monoclonal antibody specifically detects CD22 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD22 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P35329 | |
CD22 | |
Mouse myeloma cell line NS0-derived recombinant mouse Siglec-2/CD22 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
308501 | |
Unconjugated | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Rat | |
Protein A/G | |
RUO | |
12483 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |