missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Monoclonal Antibody (MEM-01), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119642
2596.79 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
CD22 Monoclonal antibody specifically detects CD22 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD22 | |
Monoclonal | |
PE | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
933 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-01 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P20273 | |
CD22 | |
Burkitt's lymphoma cell line Raji. | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |