Learn More
IGF2R Monoclonal Antibody (MEM-238), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110148
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal compartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.
IGF2R Monoclonal antibody specifically detects IGF2R in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
IGF2R | |
Monoclonal | |
PE | |
IGF2R | |
300 kDa mannose 6-phosphate receptor; AI661837; cation-independent mannose 6-phosphate receptor; cation-independent mannose 6-phosphate receptor protein; cation-independent mannose-6-phosphate receptor; cation-independent MPR; CD222; CI Man-6-P receptor; CI-M6PR; CIMPR; CI-MPR; Igf2r; IGF-II receptor; IGF-II/CI-MPR; Igfr2; insulin like growth factor 2 receptor; insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; insulin-like growth factor receptor type 2; M6P; M6P/IGF2 receptor; M6P/IGF2R; M6PR; M6P-R; mannose-6-phosphate receptor, cation independent; mannose-6-phosphate/insulin-like growth factor II receptor; MPR 300; MPR1; MPR300; MPRI; similar to mannose-6-phosphate/insulin-like growth factor II receptor | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3482 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-238 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P11717 | |
IGF2R | |
Recombinant Vaccinia virus encoding CD222. | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |