Learn More
CD3z Monoclonal Antibody (4B10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515608
Additional Details : Weight : 0.01000kg
Description
MA5-15608 targets CD247 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15608 immunogen is purified recombinant fragment of human CD247 expressed in E. Coli. MA5-15608 detects CD247 which has a predicted molecular weight of approximately 18kDa.
CD3z Monoclonal antibody specifically detects CD3z in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
CD3z | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P20963 | |
CD247 | |
Purified recombinant fragment of human CD247 expressed in E. Coli. | |
RUO | |
919 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
4B10 | |
Unconjugated | |
CD247 | |
4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |