Learn More
BAFF Monoclonal Antibody (2B), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182774
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 0.5 mL of distilled water. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. Mouse anti Human CD257 antibody, clone 2B recognizes CD257, also known as BAFF/BLyS.
BAFF Monoclonal antibody specifically detects BAFF in Human samples. It is validated for ELISA.Specifications
BAFF | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Q9Y275 | |
TNFSF13B | |
Recombinant full length mature human CD257 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
2B | |
Unconjugated | |
TNFSF13B | |
ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4 | |
Mouse | |
Protein G | |
RUO | |
10673 | |
-20°C | |
Lyophilized |