Learn More
CD26 Monoclonal Antibody (CC69), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183888
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Bovine CD26 antibody, clone CC69 recognizes bovine Dipeptidyl peptidase 4, also known as CD26, WC10, Adenosine deaminase complexing protein or Activation molecule 3 (Lee et al. 2001).
CD26 Monoclonal antibody specifically detects CD26 in Bovine, Ovine samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD26 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P81425 | |
DPP4 | |
Proprietary Immunogen. | |
250 μg | |
Primary | |
Bovine, Ovine | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
CC69 | |
Unconjugated | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Mouse | |
Protein G | |
RUO | |
100170324, 281122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |