Learn More
CD26 Monoclonal Antibody (OX-61), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170030
Additional Details : Weight : 0.01000kg
Description
MA1-70030 detects CD26 in rat samples. MA1-70030 has been successfully used in flow cytometry, immunoprecipitation, immunohistochemistry (frozen sections), and Western blot procedures. The MA1-70030 immunogen is thoracic duct lymphocytes from a mesenteric lymphodectomised rat.
CD26 Monoclonal antibody specifically detects CD26 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD26 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P14740 | |
DPP4 | |
Thoracic duct lymphocytes from a mesenteric lymphodectomised rat. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
OX-61 | |
Unconjugated | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Mouse | |
Protein G | |
RUO | |
25253 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |