missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Monoclonal Antibody (LT27), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119647
Additional Details : Weight : 0.01000kg
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.Specifications
CD27 | |
Monoclonal | |
PE | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
939 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
LT27 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P26842 | |
CD27 | |
Human peripheral blood lymphocytes. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |