missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD276, Clone: 4C4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00080381-M50.100ug
Additional Details : Weight : 0.02000kg
Specifications
CD276 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC062581.1 | |
CD276 | |
CD276 (AAH62581.1, 28 a.a. ∼ 238 a.a) partial recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
4C4 | |
Mouse monoclonal antibody raised against a partial recombinant CD276. | |
CD276 | |
B7-H3/B7H3 | |
Mouse | |
Affinity chromatography | |
RUO | |
80381 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
For Research Use Only