Läs mer
CD28 Mouse anti-Human, BB515, Clone: CD28.2, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 564493
Produktkod. 15893679
  • ONLINE EXCLUSIVE PRICE
    1270.00 SEK
    1269.02 SEK / Varje
    Save 0.98 SEK
    0.08% Av
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
CD28
Monoclonal
Brilliant Blue 515 (BB515)
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
CD28.2
CD28 antigen; T44; Tp44; TP44
Human CD28 Transfected Cell Line
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer