missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Monoclonal Antibody (CD28.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110166
Additional Details : Weight : 0.01000kg
Description
Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test. Functional Application: Costimulation.
CD28 Monoclonal antibody specifically detects CD28 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD28 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P10747 | |
Cd28 | |
DC28.1.3.3 murine T cell hybridoma transfected with human CD28 cDNA. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 κ |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
CD28.2 | |
Unconjugated | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Mouse | |
Protein A | |
RUO | |
940 | |
4° C, do not freeze | |
Liquid |