missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Monoclonal Antibody (11G7), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517723
Additional Details : Weight : 0.01000kg
Description
MA5-17723 targets CD282/TLR2/Toll-like Receptor 2 in FACS applications and shows reactivity with Human samples. The MA5-17723 immunogen is human TLR2.
TLR2 Monoclonal antibody specifically detects TLR2 in Human samples. It is validated for Flow Cytometry.Specifications
TLR2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O60603 | |
Tlr2 | |
Human TLR2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
11G7 | |
FITC | |
Tlr2 | |
CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a | |
Mouse | |
Protein G | |
RUO | |
7097 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |