missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300a Monoclonal Antibody (MEM-260), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119768
Additional Details : Weight : 0.01000kg
Description
CD300a (CMRF-35H, IRp60) is a non-MHC-specific inhibitory receptor of immunoglobulin superfamily, which contains three immunoreceptor tyrosine-based inhibitory motifs (ITIMs) that associate with SH2-containing phosphatases SHP-1 and SHP-2. CD300a is expressed on many cell types including T cells, NK cells, neutrophils, eosinophils or mast cells. Its triggering inhibits activating signals such as those of IL5, GM-CSF or eotaxin, as well as supresses mast cell degranulation or NK cell cytotoxic activity.Specifications
CD300a | |
Monoclonal | |
PE | |
Q9UGN4 | |
CD300A | |
HPB human acute lymphoid leukemia cell line. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
MEM-260 | |
CD300A | |
CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, CMRF35H, CMRF35H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60, CD300 antigen-like family member A, CD300a antigen, CMRF35-like molecule 8, IGSF12 | |
Mouse | |
Purified | |
RUO | |
11314 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |