missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300c Monoclonal Antibody (2A10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA521179
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: GYFPLSHPMT VAGPVGGSLS VQCRYEKEHR TLNKFWCRPP QILRCDKIVE TKGSAGKRNG RVSIRDSPAN LSFTVTLENL TEEDAGTYWC GVDTPWLRDF HDPIVEVEVS
CD300c Monoclonal antibody specifically detects CD300c in Human samples. It is validated for ELISA.Specifications
CD300c | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q08708 | |
CD300C | |
CD300C (NP_006669, 21 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2A10 | |
Unconjugated | |
CD300C | |
CD300 antigen-like family member C; CD300c; CD300c antigen; CD300c molecule; Clm6; CLM-6; CMRF35; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35A; CMRF-35A; CMRF35A leukocyte immunoglobulin-like receptor; CMRF35A1; CMRF35-A1; CMRF-35-like molecule 6; CMRF35-like molecule 6; Gcom1; IgSF16; immunoglobulin superfamily member 16; LIR; RGD1565161 | |
Mouse | |
Affinity chromatography | |
RUO | |
10871 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |