missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC4C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSD00005W
Additional Details : Weight : 1.24740kg
Description
Reconstitute with 100 μL of distilled water.
Specifications
CLEC4C | |
Polyclonal | |
whole serum with no preservative | |
Q8WTT0 | |
CLEC4C | |
A synthetic peptide from extracellular domain of human CD303 | |
RUO | |
170482 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
CLEC4C | |
C-type lectin superfamily member 7, Blood dendritic cell antigen 2 protein, BDCA-2, Dendritic lectin, CD303 antigen, CLEC4C, BDCA2, CLECSF11, CLECSF7, DLEC, HECL, C-type lectin domain family 4 member C | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |