Learn More
CD31 Monoclonal Antibody (WM59), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182378
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD31 monoclonal antibody, clone WM59 recognizes the human CD31 antigen, a approximately 130 kDa single pass type I transmembrane glycoprotein bearing six C2 immunoglobulin domains.
CD31 Monoclonal antibody specifically detects CD31 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
P16284 | |
Pecam1 | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry | |
WM59 | |
Biotin | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
Protein A | |
RUO | |
5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |