missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514372
Additional Details : Weight : 0.01000kg
Description
CD31 Polyclonal antibody specifically detects CD31 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD31 | |
Polyclonal | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5175 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P16284 | |
Pecam1 | |
KLH conjugated synthetic peptide between 690-718 amino acids from the C-terminal region of human CD31 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |