Learn More
CD31 Monoclonal Antibody (WM59), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182380
2693.46 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD31 monoclonal antibody, clone WM59 recognizes the human CD31 antigen, a approximately 130 kDa single pass type I transmembrane glycoprotein bearing six C2 immunoglobulin domains.
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide; pH 7.4 | |
P16284 | |
Pecam1 | |
Proprietary Immunogen. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
WM59 | |
PE | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
Protein G | |
RUO | |
5175 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |