Learn More
CD34 Monoclonal Antibody (QBEND/10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516926
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with bovine, dog, rat or sheep. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is tonsil or bone marrow. This product does not require antigen retreival using heat treatment or protein digestion. However staining of paraffin-embedded, formalin-fixed tissues may be enhanced by enzyme pre-treatment using pronase or heat treatment using citrate buffer. Mouse anti Human CD34 antibody, clone QBEND/10 recognizes the human CD34 antigen, also known as Hematopoietic progenitor cell antigen CD34.
CD34 Monoclonal antibody specifically detects CD34 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.Specifications
CD34 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P28906 | |
Cd34 | |
Human endothelial cell membrane vesicles | |
200 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
QBEND/10 | |
Unconjugated | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Mouse | |
Protein A | |
RUO | |
947 | |
4° C, do not freeze | |
Liquid |