missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Monoclonal Antibody (CB38 (NL07)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110209
Additional Details : Weight : 0.01000kg
Description
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow Cytometry.Specifications
CD36 | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
P16671 | |
CD36 | |
living human myeloid cells | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgM κ |
Flow Cytometry | |
CB38 (NL07) | |
Unconjugated | |
CD36 | |
adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor | |
Mouse | |
sequential chromatography | |
RUO | |
948 | |
4° C, do not freeze | |
Liquid |