missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Monoclonal Antibody (CB38 (NL07)), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15793
Additional Details : Weight : 0.00280kg
Description
CD36 Monoclonal antibody A15793 from Invitrogen specifically detects CD36 in Human samples. It is validated for Flow Cytometry.
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow Cytometry.Specifications
CD36 | |
Monoclonal | |
PE | |
CD36 | |
adipocyte membrane protein; BDPLT10; Cd36; CD36 antigen; CD36 antigen (collagen type I receptor, thrombospondin receptor); CD36 molecule; CD36 molecule (thrombospondin receptor); CD36 protein; CHDS7; cluster determinant 36; collagen type I receptor thrombospondin receptor; collagen type I receptor, thrombospondin receptor; FAT; FAT/CD36; fatty acid translocase; fatty acid translocase/CD36; fatty acid transport protein; glycoprotein IIIb; GP3B; GP4; GPIIIB; GPIV; I79_011940; Leukocyte differentiation antigen CD36; PAS IV; PAS4; PAS-4; PAS-4 protein; PASIV; Platelet collagen receptor; Platelet glycoprotein 4; platelet glycoprotein IV; Scarb3; scavenger receptor class B, member 3; SR-B3; Thrombospondin receptor | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
948 | |
4° C | |
IgM κ |
Flow Cytometry | |
CB38 (NL07) | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
P16671 | |
CD36 | |
living human myeloid cells | |
100 Tests | |
Primary | |
Human | |
Liquid |