missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547777
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) CD39, rmCD39L2, and rmCD39L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ENTPD2 Polyclonal antibody specifically detects ENTPD2 in Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.Specifications
ENTPD2 | |
Polyclonal | |
Unconjugated | |
ENTPD2 | |
CD39 antigen-like 1; CD39L1; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; ENTPD2; NTPDase 2; NTPDase2; NTPDase-2; testicular ecto-ATPase | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
12496, 64467 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35795, O55026 | |
ENTPD2 | |
CHO-derived recombinant mouse CD39L1/ENTPD2 Cys26-Ser462 | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |