missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 Monoclonal Antibody (3/23), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517852
Additional Details : Weight : 0.01000kg
Description
MA5-17852 targets CD40/Bp50 in FACS and IHC (F) applications and shows reactivity with Mouse samples. The MA5-17852 immunogen is mouse CD40.
CD40 Monoclonal antibody specifically detects CD40 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD40 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P27512 | |
CD40 | |
Mouse CD40 | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
3/23 | |
Unconjugated | |
CD40 | |
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 | |
Rat | |
Protein G | |
RUO | |
21939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |