Learn More
CD40 Monoclonal Antibody (LOB7/6), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180928
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD40 antibody, clone LOB7/6 recognizes the human CD40 cell surface antigen, a 48kDa glycoprotein expressed by B lymphocytes and weakly by some monocytes.
CD40 Monoclonal antibody specifically detects CD40 in Canine, Human samples. It is validated for Flow Cytometry.Specifications
CD40 | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
P25942, Q7YRL5 | |
CD40 | |
CD40 Ig(Fc) fusion protein containing the EC region of human CD40 and Fc region of human IgG | |
100 Tests | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
LOB7/6 | |
PE | |
CD40 | |
AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 | |
Mouse | |
purified | |
RUO | |
403469, 958 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |