Learn More
CD47 Monoclonal Antibody (MEM-122), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15734
Additional Details : Weight : 0.00280kg
Description
This gene encodes a membrane protein, which is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. The encoded protein is also a receptor for the C-terminal cell binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. This gene has broad tissue distribution, and is reduced in expression on Rh erythrocytes. Four alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
CD47 | |
Monoclonal | |
FITC | |
CD47 | |
IAP, MER6, OA3, CD47 antigen, CD47 glycoprotein, Rh-related antigen, leukocyte surface antigen CD47 | |
Mouse | |
Purified | |
RUO | |
961 | |
Store at 4° C | |
Liquid |
Flow Cytometry | |
MEM-122 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q08722 | |
CD47 | |
Native purified CD47 from African Green Monkey COS-7 cells | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |