missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD48 Monoclonal Antibody (MEM-102), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119119
Additional Details : Weight : 0.01000kg
Description
CD48 Monoclonal antibody specifically detects CD48 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
CD48 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P09326 | |
Cd48 | |
Burkitt's lymphoma cell line Raji. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
MEM-102 | |
Unconjugated | |
Cd48 | |
AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
Mouse | |
Protein A | |
RUO | |
962 | |
4° C, do not freeze | |
Liquid |