Learn More
ITGA4 Monoclonal Antibody (Bu49), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181535
Additional Details : Weight : 0.01000kg
Description
The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is human tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD49d monoclonal antibody, clone Bu49 recognizes human CD49d, also known as integrin alpha 4 or VLA-4 alpha subunit. CD49d is a approximately 150 kDa single pass type I transmembrane glycoprotein.
ITGA4 Monoclonal antibody specifically detects ITGA4 in Canine, Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
ITGA4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P13612 | |
ITGA4 | |
Hyatt myeloma cell line. | |
200 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
Bu49 | |
Unconjugated | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Mouse | |
Protein A | |
RUO | |
3676, 488429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |