missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA4 Monoclonal Antibody (R1-2), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517805
Additional Details : Weight : 0.01000kg
Description
ITGA4 Monoclonal antibody specifically detects ITGA4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
ITGA4 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q00651 | |
ITGA4 | |
Peyers Patch HEV binding lymphoma line (TK1) | |
300 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen) | |
R1-2 | |
Biotin | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
Rat | |
Protein G | |
RUO | |
16401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |