Learn More
ICAM-3 Monoclonal Antibody (MEM-171), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119508
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an epitope in the D2 domain of CD50 (ICAM-3), a 120-130 kDa type I membrane protein (immunoglobulin supergene family) expressed on leukocytes, endothelial cells and Langerhans cells; it is negative on platelets and erythrocytes.
ICAM-3 Monoclonal antibody specifically detects ICAM-3 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
ICAM-3 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P32942 | |
ICAM3 | |
Human granulocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
MEM-171 | |
Biotin | |
ICAM3 | |
CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3385 | |
4° C, do not freeze | |
Liquid |