missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Monoclonal Antibody (MEM-186), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119788
Additional Details : Weight : 0.01000kg
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry.Specifications
CD7 | |
Monoclonal | |
APC | |
CD7 | |
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
924 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-186 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P09564 | |
CD7 | |
Human acute myelogenous leukaemia cell line KG-1. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |