missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Receptor Monoclonal Antibody (10F11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511441
Additional Details : Weight : 0.01000kg
Description
MA5-11441 reacts with human and mouse samples in IHC (P), FACS and Western blot applications. This antibody is not suitable for MCF-7 or NIH-3T3 cells in Western blot analysis.
Transferrin Receptor Monoclonal antibody specifically detects Transferrin Receptor in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Transferrin Receptor | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P02786, Q62351 | |
TFRC | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
10F11 | |
Unconjugated | |
TFRC | |
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
Mouse | |
RUO | |
22042, 7037 | |
4° C | |
Liquid |