Learn More
Transferrin Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527739
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, Raji, NCI-H929, K562, HL-60, NIH3T3. Predicted reactivity: Dog (81%), Cat (86%), Pig (82%), Rhesus Monkey (94%), Bovine (84%), Guinea pig (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transferrin Receptor Polyclonal antibody specifically detects Transferrin Receptor in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Transferrin Receptor | |
Polyclonal | |
Unconjugated | |
TFRC | |
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22042, 7037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.47 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P02786, Q62351 | |
TFRC | |
Recombinant fragment corresponding to a region within amino acids 169 and 407 of Human CD71 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |