missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Receptor Monoclonal Antibody (R17217.1.4), Biotin, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517921
Additional Details : Weight : 0.01000kg
Description
Transferrin Receptor Monoclonal antibody specifically detects Transferrin Receptor in Mouse samples. It is validated for Flow Cytometry.Specifications
Transferrin Receptor | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q62351 | |
TFRC | |
Mouse CD71 | |
300 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
R17217.1.4 | |
Biotin | |
TFRC | |
2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr | |
Rat | |
Protein G | |
RUO | |
22042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |