Learn More
CD8 Monoclonal Antibody (4B11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180231
2261.00 SEK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval using Tris/EDTA (pH 8) is recommended for the staining of paraffin sections. A suggested positive control for this product is human tonsil Mouse anti Human CD8 antibody, clone 4B11 recognizes the human CD8 cell surface antigen, a approximately 32 kDa glycoprotein expressed by the cytotoxic/suppressor subset of T-cells and weakly by NK cells.
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
CD8 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with <0.1% sodium azide | |
P01732, P10966 | |
Cd8a, CD8B | |
Synthetic peptide derived from the carboxy terminal region of the human CD8 alpha chain coupled to a N-terminal cysteine, with the sequence C-KSDGKPSLSARYV (amino acid range 223-235). The peptide was coupled to bovine serum albumin and keyhole limpet hemocyanin. | |
RUO | |
925, 926 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
4B11 | |
Unconjugated | |
CD8B | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |