Learn More
CD90 Monoclonal Antibody (OX-7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180651
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1. Mouse anti Rat CD90 antibody, clone OX-7 recognizes rat and CD90, also known as Thy1.
CD90 Monoclonal antibody specifically detects CD90 in Guinea Pig, Mouse, Rabbit, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD90 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P01830, P01831 | |
Thy1 | |
Rat Thy1 antigen. | |
250 μg | |
Primary | |
Guinea Pig, Mouse, Rabbit, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
OX-7 | |
Unconjugated | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Mouse | |
Protein A | |
RUO | |
100339279, 100379563, 21838, 24832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |