missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA121780
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HT29 cells.
CDK8 Polyclonal antibody specifically detects CDK8 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
CDK8 | |
Polyclonal | |
Unconjugated | |
Cdk8 | |
Cdk8; CDK8 protein kinase; cell division protein kinase 8; Cell division protein kinase 8-like protein; cyclin dependent kinase 8; cyclin-dependent kinase 8; K35; mediator complex subunit CDK8; Mediator of RNA polymerase II transcription subunit CDK8; MGC126074; MGC126075; protein kinase K35; RGD1560888; wu:fk95h11 | |
Rabbit | |
Protein A | |
RUO | |
1024, 264064, 498140 | |
4° C, do not freeze | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
10mM PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P49336, Q8R3L8 | |
Cdk8 | |
Synthetic peptide: C-SQQPPQYSHQTHRY, corresponding to amino acids 451-464 of Human Cdk8. | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |