Learn More
ST6GAL1 Monoclonal Antibody (LN1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511900
Additional Details : Weight : 0.01000kg
Description
MA5-11900 targets CDw75 in FACS, IF, WB and IHC (P) applications and shows reactivity with Human and Mouse samples. Antibody does not react with rat. The molecular weight of the antigen is 53 & 87 kDa. The MA5-11900 immunogen is nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes.
ST6GAL1 Monoclonal antibody specifically detects ST6GAL1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ST6GAL1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P15907, Q64685 | |
ST6GAL1 | |
Nuclei from pokeweed mitogen-stimulated peripheral blood lymphocytes | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgM κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
LN1 | |
Unconjugated | |
ST6GAL1 | |
Alpha 2,6-ST 1; AW742324; B-cell antigen CD75; beta galactoside alpha 2,6 sialyltransferase 1; beta-galactoside alpha-2,6-sialyltransferase 1; CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1; Sialyltransferase 1; sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase); Sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase); Sialyltransferase 1 (beta-galactoside alpha-26-sialytransferase); SIAT1; ST6 beta-galactosamide alpha-2,6-sialyltranferase 1; ST6 beta-galactoside alpha-2,6-sialyltransferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; St6gal; ST6Gal I; St6gal1; ST6GalI; St6Gal-I; ST6N | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
20440, 6480 | |
4° C | |
Liquid |