Learn More
Creatine Kinase BB Monoclonal Antibody (BDI937), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191183
Additional Details : Weight : 0.01000kg
Description
MA1-91183 detects CKB from human samples. MA1-91183 has been successfully used in ELISA applications. The MA1-91183 immunogen is: Purified Human Creatine Kinase MB Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.Specifications
Creatine Kinase BB | |
Monoclonal | |
5.63 mg/mL | |
CKB | |
B-CK, CKBB | |
Mouse | |
Protein G | |
RUO | |
1152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
BDI937 | |
Unconjugated | |
P12277 | |
CKB | |
Purified Human Creatine Kinase MB | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |