missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium tetani Tetanus Toxin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA533146
Additional Details : Weight : 0.01000kg
Description
Goat anti tetanus exotoxin antibody recognizes tetanus toxin, also known as tentoxylysin, produced by the soil micro-organism Clostridium tetani.
Clostridium tetani Tetanus Toxin Polyclonal antibody specifically detects Clostridium tetani Tetanus Toxin in Bacteria samples. It is validated for ELISA, Western Blot.Specifications
Clostridium tetani Tetanus Toxin | |
Polyclonal | |
Unconjugated | |
C. tetani; Tetanus Toxin | |
Tetanus Exotoxin | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 0.09% sodium azide | |
Goat | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |