Läs mer
Clostridium tetani Tetanus Toxin Mouse anti-Bacteria, Clone: TetH6, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA174231
Beskrivning
MA1-74231 has toxin neutralizing activity. Removal of sodium azide is recommended prior to use in functional assays. Mouse anti Clostridium tetani Tetanus Toxin antibody, clone TetH6 recognizes the tetanus toxin, a neurotoxin produced by the vegetative spore of Clostridium tetani.
Tetanus toxin is a neurotoxin produced by the vegetative spore of Clostridium tetani. The toxin causes tetanus, a disease that is characterized by prolonged contraction of skeletal muscle. World-wide, approximately 1 million cases of tetanus occurs annually, resulting in approximately 300,000 to 500,000 deaths, mostly in developing countries. The tetanus toxin has a molecular weight of approximately 150kDa and consists of a heavy chain (B-chain, 100kDa) and a light chain (A-chain, 50kDa) that are connected by a disulfide bond. The toxin acts by inhibiting neurotransmitter release. It binds to peripheral neuronal synapses, is internalized and moves by retrograde transport up the axon into the spinal cord where it can move between postsynaptic and presynaptic neurons. It inhibits the release of the neurotransmitters GABA and glycine by synaptobrevin-2. This results in overactivity in the muscle. The binding of the toxin to neurons is irreversible.
Specifikationer
| Clostridium tetani Tetanus Toxin | |
| Monoclonal | |
| 1 mg/mL | |
| C. tetani; Tetanus Toxin | |
| Supernatant from Clostridium tetani. | |
| 200 μg | |
| Primary | |
| 4°C or -20°C if preferred | |
| Liquid |
| ELISA, Functional Assay, Western Blot | |
| TetH6 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.