Learn More
CNDP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544714
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ALEAYQKTGQ EIPVNVRFCL EGMEESGSEG LDELIFARKD TFFKDVDYVC Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 77%
CNDP2 Polyclonal antibody specifically detects CNDP2 in Human samples. It is validated for Western Blot.Specifications
CNDP2 | |
Polyclonal | |
Unconjugated | |
Cndp2 | |
0610010E05Rik; C76600; carnosine dipeptidase 2; carnosine dipeptidase II; CN2; CNDP dipeptidase 2; CNDP dipeptidase 2 (metallopeptidase M20 family); Cndp2; CPGL; cytosolic nonspecific dipeptidase; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase 2; Dip-2; Epididymis secretory protein Li 13; glutamate carboxypeptidase-like protein 1; HEL-S-13; HsT2298; Pep1; Pep-1; PEPA; peptidase 1; peptidase A; wu:fd20d11; wu:fe17f09; zgc:92569 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55748 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96KP4 | |
Cndp2 | |
Synthetic peptide directed towards the middle region of human CNDP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |