missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA128530
Additional Details : Weight : 0.01000kg
Descripción
PA1-28530 detects Collagen I from bovine, human samples.
Collagen I Polyclonal antibody specifically detects Collagen I in Bovine, Human, Mouse, Rat samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Especificaciones
Collagen I | |
Polyclonal | |
Biotin | |
COL1A2 | |
AA960264; AI325291; alpha 1 type I collagen; alpha 2 type I collagen; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 collagen (I); alpha-1 type 1 collagen; alpha-1 type I collagen; alpha-1 type-1 collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; alpha2(I) collagen; COL1; Col1a1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; COLA; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen; collagen 1 alpha 2 chain; collagen a2; collagen alpha 1 chain type I; collagen alpha 2(I) chain precursor; collagen alpha-1 chain; collagen alpha-1 chain precursor; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tend | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1277, 1278, 12842, 12843, 282187, 282188, 29393, 84352 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.125M sodium borate with 0.075M NaCl, 10mg/mL BSA, 5mM EDTA and 0.01% sodium azide; pH 8.0 | |
P02452, P02453, P02454, P02465, P02466, P08123, P11087, Q01149 | |
COL1A1, COL1A2 | |
Collagen Type I from adult human and bovine tissues. | |
50 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |