missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539184
Additional Details : Weight : 0.00900kg
Description
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
Connexin 47 | |
Polyclonal | |
Unconjugated | |
GJC2 | |
Gap junction gamma-2 protein, Connexin-46.6, Cx46.6, Cx47, Gap junction alpha-12 protein, GJC2, GJA12, CXG2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118454, 497913, 57165 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5T442, Q8BQU6, Q80XF7 | |
GJC2 | |
A synthetic peptide derived from the internal region of human CXG2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |