missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin 3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518276
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and porcine based on sequence homology.
Coronin 3 Polyclonal antibody specifically detects Coronin 3 in Human samples. It is validated for Western Blot.Specifications
Coronin 3 | |
Polyclonal | |
Unconjugated | |
Coro1c | |
AL022675; AW455561; AW548837; Coro1c; coronin 1c; coronin 3; coronin, actin binding protein 1C; coronin, actin binding protein, 1C; coronin, actin-binding protein, 1C; coronin-1C; Coronin-3; CRN2; CRNN4; hCRNN4; RGD1564490 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23603 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9ULV4 | |
Coro1c | |
Synthetic peptide sequence (RISKLEQQMAKIAA) corresponding to the C-terminus amino acids of CORO1C | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |