missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nkx2.5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519138
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and canine based on sequence homology.
Nkx2.5 Polyclonal antibody specifically detects Nkx2.5 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Nkx2.5 | |
Polyclonal | |
Unconjugated | |
Nkx2-5 | |
Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
114109, 1482, 18091 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O35767, P42582, P52952 | |
Nkx2-5 | |
Synthetic peptide sequence (PRAYSDPDPAKDPR) corresponding to the internal amino acids of NKX2-5 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |