Läs mer
Anti-Dechlorane plus, TRC
Produktkod. 30362643
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30362643 25 mg
1 options
Produktkod. 30362643 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22663525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dechlorane Plus Anti
CAS 135821-74-8
Molekylformel C18 H12 Cl12
Kvantitet 25 mg
Synonym anti-Dechlorane A, 1,4:7,10-Dimethanodibenzo[a,e]cyclooctene, 1,2,3,4,7,8,9,10,13,13,14,14-dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-, (1α,4α,4aβ,6aα,7β,10β,10aα,12aβ)- (9CI), rel-(1R,4S,4aS,6aS,7S,10R,10aR,12aR)-1,2,3,4,7,8,9,10,13,13,14,14-Dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-1,4:7,10-dimethanodibenzo[a,e]cyclooctene, anti-DP, anti-Dechlorane plus, anti-Dodecachloropentacyclooctadecadiene, 1,4:7,10-Dimethanodibenzo[a,e]cyclooctene, 1,2,3,4,7,8,9,10,13,13,14,14-dodecachloro-1,4,4a,5,6,6a,7,10,10a,11,12,12a-dodecahydro-, (1R,4S,4aS,6aS,7S,10R,10aR,12aR)-rel-
InChI formel InChI=1S/C18H12Cl12/c19-9-10(20)15(25)7-3-4-8-6(2-1-5(7)13(9,23)17(15,27)28)14(24)11(21)12(22)16(8,26)18(14,29)30/h5-8H,1-4H2/t5-,6-,7+,8+,13?,14?,15?,16?
LEDER ClC1=C(Cl)C2(Cl)[C@H]3CC[C@H]4[C@@H](CC[C@H]3C1(Cl)C2(Cl)Cl)C5(Cl)C(=C(Cl)C4(Cl)C5(Cl)Cl)Cl
IUPAC-namn (1R,2R,5R,6R,9S,10S,13S,14S)-1,6,7,8,9,14,15,16,17,17,18,18-dodecachloropentacyclo[12.2.1.16,9.02,13.05,10]octadeca-7,15-diene
Molekylvikt (g/mol) 653.72
Formel vikt 647.72